Assay Details
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129170.6 | XP_003129218.4 | 657 | 75 | ||
XM_021100663.1 | XP_020956322.1 | 591 | 75 | |||
GenBank mRNA | AK344319.1 | - | 5 - 6 | 558 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
