Assay Details
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927860.5 | XP_001927895.1 | 157 | 107 | ||
XM_005663525.3 | XP_005663582.1 | 157 | 107 | |||
GenBank mRNA | AK344348.1 | - | 1 - 2 | 89 | 107 | |
AY610243.1 | - | 1 - 2 | 69 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
