Assay Details
Gene Symbol: RAB2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013978245.2 | XP_013833699.1 | 603 | 57 | ||
XM_013978246.2 | XP_013833700.1 | 578 | 57 | |||
GenBank mRNA | AK344438.1 | - | 4 - 5 | 310 | 57 | |
AK389764.1 | - | 4 - 5 | 285 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
