Assay Details
| Gene Symbol: LOC100524972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122664.4 | XP_003122712.2 | 352 | 121 | ||
| GenBank mRNA | AK344539.1 | - | 2 - 3 | 313 | 121 | |
More Information
Set Membership: |
|

LOC100524972
Genomic Map