Assay Details
Gene Symbol: LOC100520923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021101772.1 | XP_020957431.1 | 2605 | 92 | ||
XM_021101774.1 | XP_020957433.1 | 1351 | 92 | |||
XM_021101775.1 | XP_020957434.1 | 1270 | 92 | |||
GenBank mRNA | AK344700.1 | - | 5 - 6 | 803 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
