Assay Details
| Gene Symbol: PRSS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655190.3 | XP_005655247.1 | 6497 | 85 | ||
| XM_005655191.3 | XP_005655248.1 | 6381 | 85 | |||
| XM_021086746.1 | XP_020942405.1 | 6381 | 85 | |||
| XM_021086747.1 | XP_020942406.1 | 6497 | 85 | |||
| GenBank mRNA | AK345043.1 | - | 2 - 3 | 131 | 85 | |
| AK396363.1 | - | 2 - 3 | 155 | 85 | ||
More Information
Set Membership: |
|

PRSS22
Genomic Map