Assay Details
| Gene Symbol: PRSS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655190.3 | XP_005655247.1 | 6680 | 100 | ||
| XM_005655191.3 | XP_005655248.1 | 6564 | 100 | |||
| GenBank mRNA | AK345043.1 | - | 3 - 4 | 314 | 100 | |
| AK396363.1 | - | 3 - 4 | 338 | 100 | ||
More Information
Set Membership: |
|

PRSS22
Genomic Map