Assay Details
Gene Symbol: SDF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003356108.4 | XP_003356156.1 | 953 | 60 | ||
XM_021097430.1 | XP_020953089.1 | 881 | 60 | |||
GenBank mRNA | AK345230.1 | - | 4 - 5 | 1204 | 60 | |
AK351231.1 | - | 5 - 6 | 842 | 60 | ||
AK351959.1 | - | 5 - 6 | 842 | 60 | ||
AK389368.1 | - | 5 - 6 | 860 | 60 | ||
AK399152.1 | - | 5 - 6 | 842 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
