Assay Details
| Gene Symbol: NIPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075601.1 | XP_020931260.1 | 954 | 97 | ||
| XM_021075602.1 | XP_020931261.1 | 826 | 97 | |||
| XM_021075603.1 | XP_020931262.1 | 837 | 97 | |||
| XM_021075604.1 | XP_020931263.1 | 692 | 97 | |||
| XM_021075605.1 | XP_020931264.1 | 574 | 97 | |||
| GenBank mRNA | AK345307.1 | - | 6 - 7 | 939 | 97 | |
More Information
Set Membership: |
|

NIPA2
Genomic Map