Assay Details
| Gene Symbol: SLC35A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986120.2 | XP_013841574.2 | 1210 | 56 | ||
| XM_021079677.1 | XP_020935336.1 | 1026 | 56 | |||
| XM_021079679.1 | XP_020935338.1 | 620 | 56 | |||
| GenBank mRNA | AK345340.1 | - | 4 - 5 | 1179 | 56 | |
More Information
Set Membership: |
|

SLC35A2
Genomic Map