Assay Details
Gene Symbol: SCIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003358218.4 | XP_003358266.1 | 1277 | 83 | ||
GenBank mRNA | AK345456.1 | - | 3 - 4 | 431 | 83 | |
AK345557.1 | - | 3 - 4 | 464 | 83 | ||
AK350290.1 | - | 3 - 4 | 415 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
