Assay Details
Gene Symbol: LOC100520512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068089.1 | XP_020923748.1 | 8217 | 90 | ||
XM_021068090.1 | XP_020923749.1 | 8786 | 90 | |||
XM_021068091.1 | XP_020923750.1 | 8005 | 90 | |||
XM_021068092.1 | XP_020923751.1 | 8054 | 90 | |||
XM_021068093.1 | XP_020923752.1 | 8053 | 90 | |||
XM_021068094.1 | XP_020923753.1 | 4915 | 90 | |||
XM_021068095.1 | XP_020923754.1 | 7941 | 90 | |||
XM_021068096.1 | XP_020923755.1 | 8302 | 90 | |||
XM_021068097.1 | XP_020923756.1 | 7940 | 90 | |||
XM_021068098.1 | XP_020923757.1 | 7949 | 90 | |||
XM_021068099.1 | XP_020923758.1 | 7948 | 90 | |||
XM_021068100.1 | XP_020923759.1 | 7911 | 90 | |||
XM_021068101.1 | XP_020923760.1 | 7909 | 90 | |||
GenBank mRNA | AK345887.1 | - | 12 - 13 | 1269 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
