Assay Details
Gene Symbol: LOC100520512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068089.1 | XP_020923748.1 | 8350 | 93 | ||
XM_021068090.1 | XP_020923749.1 | 8919 | 93 | |||
XM_021068091.1 | XP_020923750.1 | 8138 | 93 | |||
XM_021068092.1 | XP_020923751.1 | 8241 | 93 | |||
XM_021068093.1 | XP_020923752.1 | 8186 | 93 | |||
XM_021068094.1 | XP_020923753.1 | 5102 | 93 | |||
XM_021068095.1 | XP_020923754.1 | 8128 | 93 | |||
XM_021068096.1 | XP_020923755.1 | 8435 | 93 | |||
XM_021068097.1 | XP_020923756.1 | 8073 | 93 | |||
XM_021068098.1 | XP_020923757.1 | 8136 | 93 | |||
XM_021068099.1 | XP_020923758.1 | 8081 | 93 | |||
XM_021068100.1 | XP_020923759.1 | 8098 | 93 | |||
XM_021068101.1 | XP_020923760.1 | 8042 | 93 | |||
GenBank mRNA | AK345887.1 | - | 14 - 15 | 1402 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
