Assay Details
Gene Symbol: LOC100520512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068089.1 | XP_020923748.1 | 6969 | 129 | ||
XM_021068090.1 | XP_020923749.1 | 7538 | 129 | |||
XM_021068091.1 | XP_020923750.1 | 6757 | 129 | |||
XM_021068092.1 | XP_020923751.1 | 6968 | 129 | |||
XM_021068093.1 | XP_020923752.1 | 6967 | 129 | |||
XM_021068094.1 | XP_020923753.1 | 3829 | 129 | |||
XM_021068095.1 | XP_020923754.1 | 6855 | 129 | |||
XM_021068096.1 | XP_020923755.1 | 7216 | 129 | |||
XM_021068097.1 | XP_020923756.1 | 6854 | 129 | |||
XM_021068098.1 | XP_020923757.1 | 6863 | 129 | |||
XM_021068099.1 | XP_020923758.1 | 6862 | 129 | |||
XM_021068100.1 | XP_020923759.1 | 6825 | 129 | |||
XM_021068101.1 | XP_020923760.1 | 6823 | 129 | |||
GenBank mRNA | AK345887.1 | - | 2 - 3 | 183 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
