Assay Details
Gene Symbol: LOC100520512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068089.1 | XP_020923748.1 | 7033 | 94 | ||
XM_021068090.1 | XP_020923749.1 | 7602 | 94 | |||
XM_021068091.1 | XP_020923750.1 | 6821 | 94 | |||
XM_021068092.1 | XP_020923751.1 | 7032 | 94 | |||
XM_021068093.1 | XP_020923752.1 | 7031 | 94 | |||
XM_021068094.1 | XP_020923753.1 | 3893 | 94 | |||
XM_021068095.1 | XP_020923754.1 | 6919 | 94 | |||
XM_021068096.1 | XP_020923755.1 | 7280 | 94 | |||
XM_021068097.1 | XP_020923756.1 | 6918 | 94 | |||
XM_021068098.1 | XP_020923757.1 | 6927 | 94 | |||
XM_021068099.1 | XP_020923758.1 | 6926 | 94 | |||
XM_021068100.1 | XP_020923759.1 | 6889 | 94 | |||
XM_021068101.1 | XP_020923760.1 | 6887 | 94 | |||
GenBank mRNA | AK345887.1 | - | 3 - 4 | 247 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
