Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003355210.4 | XP_003355258.1 | 182 | 105 | ||
XM_021089932.1 | XP_020945591.1 | 181 | 105 | |||
GenBank mRNA | AK346152.1 | - | 1 - 2 | 133 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003355210.4, XM_021089932.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
