Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003355210.4 | XP_003355258.1 | 103 | |||
XM_005663519.3 | XP_005663576.2 | 103 | ||||
XM_013997208.2 | XP_013852662.2 | 103 | ||||
XM_021089931.1 | XP_020945590.1 | 103 | ||||
GenBank mRNA | AK346152.1 | - | 2 - 3 | 705 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
