Assay Details
Gene Symbol: COLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003361689.3 | XP_003361737.2 | 516 | 60 | ||
XM_013986941.2 | XP_013842395.2 | 406 | 60 | |||
XM_013986942.2 | XP_013842396.2 | 334 | 60 | |||
XM_021090562.1 | XP_020946221.1 | 1858 | 60 | |||
XM_021090563.1 | XP_020946222.1 | 1607 | 60 | |||
XM_021090564.1 | XP_020946223.1 | 1687 | 60 | |||
XM_021090565.1 | XP_020946224.1 | 1699 | 60 | |||
XM_021090566.1 | XP_020946225.1 | 1459 | 60 | |||
XM_021090567.1 | XP_020946226.1 | 1976 | 60 | |||
XM_021090568.1 | XP_020946227.1 | 1952 | 60 | |||
XM_021090569.1 | XP_020946228.1 | 1771 | 60 | |||
XM_021090570.1 | XP_020946229.1 | 1960 | 60 | |||
XM_021090571.1 | XP_020946230.1 | 1850 | 60 | |||
XM_021090572.1 | XP_020946231.1 | 1788 | 60 | |||
XM_021090573.1 | XP_020946232.1 | 1763 | 60 | |||
XM_021090574.1 | XP_020946233.1 | 1446 | 60 | |||
XM_021090575.1 | XP_020946234.1 | 1510 | 60 | |||
GenBank mRNA | AK346453.1 | - | 4 - 5 | 384 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
