Assay Details
Gene Symbol: ATP5MPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003128756.5 | XP_003128804.1 | 279 | 102 | ||
GenBank mRNA | AK346640.1 | - | 1 | 212 | 102 | |
AK392272.1 | - | 1 | 244 | 102 | ||
AK399706.1 | - | 1 | 240 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
