Assay Details
Gene Symbol: POP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127004.4 | XP_003127052.1 | 82 | |||
GenBank mRNA | AK346866.1 | - | 3 - 4 | 505 | 82 | |
AK350369.1 | - | 3 - 4 | 82 | |||
AY609682.1 | - | 3 - 4 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
