Assay Details
| Gene Symbol: NDUFS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672445.3 | XP_005672502.2 | 146 | 98 | ||
| XM_021077054.1 | XP_020932713.1 | 146 | 98 | |||
| XM_021077055.1 | XP_020932714.1 | 146 | 98 | |||
| GenBank mRNA | AK347217.1 | - | 1 - 2 | 106 | 98 | |
| AY610394.1 | - | 1 - 2 | 99 | 98 | ||
| JN703390.1 | - | 1 - 2 | 99 | 98 | ||
More Information
Set Membership: |
|

NDUFS4
Genomic Map