Assay Details
| Gene Symbol: PPP1R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360157.4 | XP_003360205.2 | 5791 | 95 | ||
| GenBank mRNA | AK347238.1 | - | 1 | 1022 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R3A| Gene Symbol: PPP1R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360157.4 | XP_003360205.2 | 5791 | 95 | ||
| GenBank mRNA | AK347238.1 | - | 1 | 1022 | 95 | |
Set Membership: |
|