Assay Details
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021079223.1 | XP_020934882.1 | 1016 | 105 | ||
XM_021079224.1 | XP_020934883.1 | 1016 | 105 | |||
XM_021079225.1 | XP_020934884.1 | 1017 | 105 | |||
XM_021079226.1 | XP_020934885.1 | 980 | 105 | |||
XM_021079227.1 | XP_020934886.1 | 389 | 105 | |||
GenBank mRNA | AK347335.1 | - | 3 - 4 | 536 | 105 | |
AK347434.1 | - | 4 - 5 | 682 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
