Assay Details
| Gene Symbol: FAM181B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129715.3 | XP_003129763.2 | 1417 | 107 | ||
| GenBank mRNA | AK347595.1 | - | 1 | 1421 | 107 | |
| AK392183.1 | - | 1 | 1423 | 107 | ||
More Information
Set Membership: |
|

FAM181B
Genomic Map