Assay Details
| Gene Symbol: SPATA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077958.1 | XP_020933617.1 | 4699 | 93 | ||
| XM_021077959.1 | XP_020933618.1 | 806 | 93 | |||
| XM_021077960.1 | XP_020933619.1 | 964 | 93 | |||
| GenBank mRNA | AK348246.1 | - | 2 - 3 | 564 | 93 | |
More Information
Set Membership: |
|

SPATA2
Genomic Map