Assay Details
Gene Symbol: TMA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129012.4 | XP_003129060.1 | 379 | 60 | ||
XM_021100589.1 | XP_020956248.1 | 381 | 60 | |||
GenBank mRNA | AK348580.1 | - | 4 - 5 | 308 | 60 | |
AK395459.1 | - | 4 - 5 | 335 | 60 | ||
AK400000.1 | - | 4 - 5 | 355 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
