Assay Details
| Gene Symbol: PPP1R36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926455.5 | XP_001926490.2 | 1353 | 73 | ||
| XM_021099928.1 | XP_020955587.1 | 1260 | 73 | |||
| XM_021099929.1 | XP_020955588.1 | 1418 | 73 | |||
| GenBank mRNA | AK348587.1 | - | 8 - 9 | 1118 | 73 | |
More Information
Set Membership: |
|

PPP1R36
Genomic Map