Assay Details
| Gene Symbol: RFC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670675.3 | XP_005670732.1 | 803 | 66 | ||
| GenBank mRNA | AK348613.1 | - | 7 - 8 | 789 | 66 | |
| AK399681.1 | - | 7 - 8 | 777 | 66 | ||
More Information
Set Membership: |
|

RFC5
Genomic Map