Assay Details
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003357318.4 | XP_003357366.1 | 821 | 65 | ||
GenBank mRNA | AK348707.1 | - | 4 - 5 | 776 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
