Assay Details
| Gene Symbol: LOC100511846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084518.1 | XP_020940177.1 | 113 | |||
| GenBank mRNA | AK349071.1 | - | 1 - 2 | 535 | 113 | |
More Information
Set Membership: |
|

LOC100511846
Genomic Map