Assay Details
| Gene Symbol: TRIM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067910.1 | XP_020923569.1 | 2455 | 54 | ||
| GenBank mRNA | AK349146.1 | - | 6 - 7 | 1173 | 54 | |
More Information
Set Membership: |
|

TRIM16
Genomic Map