Assay Details
Gene Symbol: LOC102162486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005657738.3 | XP_005657795.1 | 462 | 85 | ||
XM_021078854.1 | XP_020934513.1 | 471 | 85 | |||
GenBank mRNA | AK349205.1 | - | 1 - 2 | 380 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
