Assay Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021065993.1 | XP_020921652.1 | 1472 | 72 | ||
XM_021065994.1 | XP_020921653.1 | 1472 | 72 | |||
XM_021065995.1 | XP_020921654.1 | 1474 | 72 | |||
XM_021065996.1 | XP_020921655.1 | 1473 | 72 | |||
XM_021065997.1 | XP_020921656.1 | 287 | 72 | |||
XM_021065998.1 | XP_020921657.1 | 288 | 72 | |||
XM_021065999.1 | XP_020921658.1 | 1475 | 72 | |||
XM_021066000.1 | XP_020921659.1 | 1476 | 72 | |||
GenBank mRNA | AK349474.1 | - | 2 - 3 | 343 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
