Assay Details
Gene Symbol: RCOR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021063868.1 | XP_020919527.1 | 1146 | 72 | ||
XM_021063869.1 | XP_020919528.1 | 1143 | 72 | |||
XM_021063870.1 | XP_020919529.1 | 1146 | 72 | |||
XM_021063871.1 | XP_020919530.1 | 1142 | 72 | |||
XM_021063872.1 | XP_020919531.1 | 1143 | 72 | |||
XM_021063873.1 | XP_020919532.1 | 1491 | 72 | |||
XM_021063874.1 | XP_020919533.1 | 1697 | 72 | |||
XM_021063875.1 | XP_020919534.1 | 1323 | 72 | |||
GenBank mRNA | AK349543.1 | - | 8 - 9 | 1108 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
