Assay Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095055.1 | XP_020950714.1 | 667 | 86 | ||
| XM_021095056.1 | XP_020950715.1 | 711 | 86 | |||
| XM_021095057.1 | XP_020950716.1 | 498 | 86 | |||
| GenBank mRNA | AK349686.1 | - | 5 - 6 | 711 | 86 | |
More Information
Set Membership: |
|

CCDC106
Genomic Map