Assay Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003481371.3 | XP_003481419.1 | 85 | |||
GenBank mRNA | AK349740.1 | - | 1 - 2 | 94 | 85 | |
AK392604.1 | - | 1 - 2 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003481371.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
