Assay Details
| Gene Symbol: TEKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127787.3 | XP_003127835.1 | 318 | 62 | ||
| XM_005665212.2 | XP_005665269.1 | 362 | 62 | |||
| GenBank mRNA | AK351015.1 | - | 2 - 3 | 235 | 62 | |
More Information
Set Membership: |
|

TEKT2
Genomic Map