Assay Details
Gene Symbol: LOC100738160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021077836.1 | XP_020933495.1 | 261 | 108 | ||
GenBank mRNA | AK351348.1 | - | 1 - 2 | 113 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
