Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125214.4 | XP_003125262.1 | 673 | 104 | ||
XM_003125215.4 | XP_003125263.1 | 437 | 104 | |||
XM_003354857.4 | XP_003354905.1 | 280 | 104 | |||
XM_021087596.1 | XP_020943255.1 | 434 | 104 | |||
XM_021087597.1 | XP_020943256.1 | 255 | 104 | |||
GenBank mRNA | AK352052.1 | - | 2 - 3 | 660 | 104 | |
AY609672.1 | - | 2 - 3 | 311 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
