Assay Details
| Gene Symbol: LOC100625149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065574.1 | XP_020921233.1 | 5164 | 122 | ||
| GenBank mRNA | AK389201.1 | - | 2 - 3 | 253 | 122 | |
More Information
Set Membership: |
|

LOC100625149
Genomic Map