Assay Details
| Gene Symbol: CDHR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132219.4 | XP_003132267.1 | 2157 | 59 | ||
| XM_021068791.1 | XP_020924450.1 | 1649 | 59 | |||
| GenBank mRNA | AK389210.1 | - | 3 - 4 | 752 | 59 | |
More Information
Set Membership: |
|

CDHR4
Genomic Map