Assay Details
| Gene Symbol: CSNK2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134359.5 | XP_003134407.1 | 3632 | 88 | ||
| GenBank mRNA | AK231567.1 | - | 1 | 2025 | 88 | |
| AK240411.1 | - | 1 | 1847 | 88 | ||
| AK393557.1 | - | 1 | 1266 | 88 | ||
More Information
Set Membership: |
|

CSNK2A1
Genomic Map