Assay Details
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003354871.4 | XP_003354919.1 | 289 | 107 | ||
XM_003354872.4 | XP_003354920.1 | 366 | 107 | |||
XM_003354874.2 | XP_003354922.1 | 346 | 107 | |||
XM_005662649.2 | XP_005662706.1 | 405 | 107 | |||
XM_021087664.1 | XP_020943323.1 | 826 | 107 | |||
GenBank mRNA | AK231697.1 | - | 2 - 3 | 275 | 107 | |
AK237036.1 | - | 2 - 3 | 225 | 107 | ||
AK391141.1 | - | 2 - 3 | 281 | 107 | ||
AY609945.1 | - | 3 - 4 | 295 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
