Assay Details
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131870.4 | XP_003131918.1 | 694 | 85 | ||
XM_005669123.3 | XP_005669180.1 | 699 | 85 | |||
XM_005669126.2 | XP_005669183.1 | 269 | 85 | |||
XM_021067695.1 | XP_020923354.1 | 389 | 85 | |||
XM_021067697.1 | XP_020923356.1 | 697 | 85 | |||
XM_021067698.1 | XP_020923357.1 | 163 | 85 | |||
GenBank mRNA | AK231717.1 | - | 2 - 3 | 680 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
