Assay Details
Gene Symbol: SEPTIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003135344.5 | XP_003135392.1 | 583 | 79 | ||
XM_005673867.3 | XP_005673924.1 | 334 | 79 | |||
XM_005673868.3 | XP_005673925.1 | 560 | 79 | |||
XM_013986371.2 | XP_013841825.2 | 475 | 79 | |||
XM_013986372.2 | XP_013841826.2 | 475 | 79 | |||
XM_013986373.2 | XP_013841827.2 | 474 | 79 | |||
XM_013986374.2 | XP_013841828.2 | 473 | 79 | |||
XM_013986377.2 | XP_013841831.1 | 560 | 79 | |||
XM_021080245.1 | XP_020935904.1 | 583 | 79 | |||
XM_021080247.1 | XP_020935906.1 | 560 | 79 | |||
XM_021080248.1 | XP_020935907.1 | 481 | 79 | |||
XM_021080249.1 | XP_020935908.1 | 560 | 79 | |||
XM_021080250.1 | XP_020935909.1 | 228 | 79 | |||
XM_021080251.1 | XP_020935910.1 | 227 | 79 | |||
XM_021080252.1 | XP_020935911.1 | 581 | 79 | |||
XM_021080253.1 | XP_020935912.1 | 254 | 79 | |||
GenBank mRNA | AK231751.1 | - | 3 - 4 | 297 | 79 | |
AK237053.1 | - | 2 - 3 | 239 | 79 | ||
AK349008.1 | - | 2 - 3 | 206 | 79 | ||
KU358538.1 | - | 2 - 3 | 291 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021080252.1, XM_021080253.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
