Assay Details
Gene Symbol: SLC22A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK231777.1 | - | 1 - 2 | 105 | 81 | |
AK232296.1 | - | 1 - 2 | 126 | 81 | ||
AK234863.1 | - | 1 - 2 | 126 | 81 | ||
AK345122.1 | - | 1 - 2 | 108 | 81 | ||
AK349481.1 | - | 1 - 2 | 320 | 81 | ||
AK389923.1 | - | 1 - 2 | 115 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
