Assay Details
Gene Symbol: CCDC127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021076939.1 | XP_020932598.1 | 124 | 112 | ||
GenBank mRNA | AK231814.1 | - | 1 - 2 | 60 | 112 | |
AK398281.1 | - | 1 - 2 | 79 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021076939.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
