Assay Details
Gene Symbol: RO60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK231823.1 | - | 1 - 2 | 69 | 89 | |
AK238342.1 | - | 2 - 3 | 745 | 89 | ||
AK390549.1 | - | 2 - 3 | 777 | 89 | ||
AK396336.1 | - | 2 - 3 | 724 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
