Assay Details
Gene Symbol: TMEM231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126861.6 | XP_003126909.4 | 241 | 79 | ||
GenBank mRNA | AK231852.1 | - | 1 - 2 | 238 | 79 | |
AK232544.1 | - | 1 - 2 | 206 | 79 | ||
AK348245.1 | - | 1 - 2 | 227 | 79 | ||
AK352234.1 | - | 1 - 2 | 206 | 79 | ||
AK397631.1 | - | 1 - 2 | 227 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
