Assay Details
| Gene Symbol: ATP6V0C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124744.4 | XP_003124792.1 | 451 | 71 | ||
| GenBank mRNA | AK231969.1 | - | 2 - 3 | 403 | 71 | |
| AK346661.1 | - | 2 - 3 | 375 | 71 | ||
| AK391769.1 | - | 2 - 3 | 429 | 71 | ||
| AY610014.1 | - | 2 - 3 | 401 | 71 | ||
More Information
Set Membership: |
|

ATP6V0C
Genomic Map